Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUVBL2 Recombinant Rabbit Monoclonal Antibody (3U4L10)
GREENER_CHOICE

Catalog No. PIMA535467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535467 Supplier Thermo Scientific Supplier No. MA535467
Add to Cart
Add to Cart

RUVBL2 Recombinant Rabbit Monoclonal Antibody (ARC0933)

RUVBL2 Recombinant Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RUVBL2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3U4L10
Concentration 0.166 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ruvbl2
Gene Accession No. Q9WTM5, Q9Y230
Gene Alias 48 kDa TATA box-binding protein-interacting protein; 48 kDa TBP-interacting protein; 51 kDa erythrocyte cytosolic protein; CGI-46; ECP51; ECP-51; erythrocyte cytosolic protein, 51-KD; INO80 complex subunit J; INO80J; liebeskummer; lik; mp47; p47; p47 protein; Repressing pontin 52; reptin; Reptin 52; reptin52 protein; RuvB (E coli homolog)-like 2; RuvB like AAA ATPase 2; RuvB like AAA ATPase 2 S homeolog; ruvbl2; RUVBL2 protein; ruvbl2.S; ruvB-like 2; RuvB-like AAA ATPase 2; RuvB-like DNA helicase reptin; RuvB-like protein 2; RVB2; TAP54-beta; TIH2; TIP48; TIP49B; TIP60-associated protein 54-beta; wu:fi25f01; XELAEV_180378471mg; xReptin; zReptin
Gene Symbols Ruvbl2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human Reptin/RUVBL2 (Q9Y230).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10856, 20174, 292907
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.