Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUVBL2 Monoclonal Antibody (OTI2B9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504384S


Description
RUVBL2 Monoclonal antibody specifically detects RUVBL2 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
RUVBL2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P70195, Q99436, Q9JHW0 | |
Mouse | |
Affinity chromatography | |
RUO | |
100686423, 19177, 5695, 85492 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI2B9 | |
Unconjugated | |
Psmb7 | |
48 kDa TATA box-binding protein-interacting protein; 48 kDa TBP-interacting protein; 51 kDa erythrocyte cytosolic protein; CGI-46; ECP51; ECP-51; erythrocyte cytosolic protein, 51-KD; INO80 complex subunit J; INO80J; liebeskummer; lik; mp47; p47; p47 protein; Repressing pontin 52; reptin; Reptin 52; reptin52 protein; RuvB (E coli homolog)-like 2; RuvB like AAA ATPase 2; RuvB like AAA ATPase 2 S homeolog; ruvbl2; RUVBL2 protein; ruvbl2.S; ruvB-like 2; RuvB-like AAA ATPase 2; RuvB-like DNA helicase reptin; RuvB-like protein 2; RVB2; TAP54-beta; TIH2; TIP48; TIP49B; TIP60-associated protein 54-beta; wu:fi25f01; XELAEV_180378471mg; xReptin; zReptin | |
Human recombinant protein fragment corresponding to amino acids 58-277 of human PSMB7 produced in E.coli. | |
30 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction