Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RuvC Antibody (RuvC 7F1/5), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250281JF646
Description
RuvC Monoclonal antibody specifically detects RuvC in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
RuvC | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Ruv C protein from E. Coli | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry | |
RuvC 7F1/5 | |
Western Blot, ELISA, Immunohistochemistry | |
Mouse | |
Protein A purified | |
DNA Repair, Epigenetics | |
E. coli | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction