Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RVF Virus (TAN/Dod-002/07) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581827
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rift Valley fever virus (TAN/Dod-002/07) RVFV-L.

Specifications
RVF Virus (TAN/Dod-002/07) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
ELISA | |
1 mg/mL | |
PBS with no preservative | |
Recombinant Rift Valley fever virus (TAN/Dod-002/07) RVFV-L Protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction