Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RWDD2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556348
Description
Immunogen sequence: SASVKESLQL QLLEMEMLFS MFPNQGEVKL EDVNALTNIK RYLEGTREAL PPKIEFVITL QIEEPKVKID LQVTMPHSYP YVALQLFGRS SELDRHQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Specifications
| RWDD2A | |
| Polyclonal | |
| Unconjugated | |
| RWDD2A | |
| 1700030C20Rik; AI848608; dJ747H23.2; RWD domain containing 2; RWD domain containing 2A; RWD domain-containing protein 2A; RWDD2; RWDD2A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 112611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UIY3 | |
| RWDD2A | |
| Recombinant protein corresponding to Human RWDD2A. Recombinant protein control fragment (Product #RP-94596). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction