Learn More
Invitrogen™ RWDD3 Polyclonal Antibody

Description
A suggested positive control is mouse kidney tissue lysate. PA5-20821 can be used with blocking peptide PEP-0935.
Specifications
Specifications
| Antigen | RWDD3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RWDD3 |
| Gene Accession No. | P0C7N0, Q8VIL2, Q9Y3V2 |
| Gene Alias | 2510027J23Rik; 3110037C01Rik; AI035684; AI662458; pituitary tumor X2CR1 protein; RSUME; RWD domain containing 3; RWD domain-containing protein 3; RWD domain-containing sumoylation enhancer; RWD-containing sumoylation enhancer; Rwdd3; X2cr1 |
| Gene Symbols | RWDD3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.