Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RWDD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114147 Supplier Invitrogen™ Supplier No. PA5114147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESLLSEPMVH ELVLWIQQNL RHILSQPETG SGSEKCTFST STTMDDGLWI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 79%; Rat: 86%.

RWDD3 (RSUME), a small RWD-containing protein, has a central role in sumoylation by enhancing SUMO conjugation in the regulatory network of immune-inflammatory signals. RWDD3 increases Ikappa-Beta sumoylation and stability. In addition, RWDD3 inhibits TNF-a-induced kappaB-LUC (Luciferase) reporter activity, showing the functional consequence of Ikappa-Beta increased stability. RSUME-enhanced sumoylation of Ikappa-Beta leads to the inhibition of NF-kappa-Beta activity on two well-known inflammatory genes, IL-8 and cyclooxygenase-2 (Cox-2) and therefore may also favor anti-inflammatory pathways. Expression of RWDD3 was induced under hypoxic conditions and it has a potential role during vascularization. Both BMP-4 and RWDD3 may be interesting targets for inhibiting steps involved in pituitary tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RWDD3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RWDD3
Gene Accession No. Q9Y3V2
Gene Alias 2510027J23Rik; 3110037C01Rik; AI035684; AI662458; pituitary tumor X2CR1 protein; RSUME; RWD domain containing 3; RWD domain-containing protein 3; RWD domain-containing sumoylation enhancer; RWD-containing sumoylation enhancer; Rwdd3; X2cr1
Gene Symbols RWDD3
Host Species Rabbit
Immunogen Immunogen sequence: ESLLSEPMVH ELVLWIQQNL RHILSQPETG SGSEKCTFST STTMDDGLWI (aa 92-141).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25950
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.