Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RWDD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557691 Supplier Invitrogen™ Supplier No. PA557691
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALRSIYEGD ESFRELSPVS FQYRIGENGD PKAFLIEISW TETYPQTPPI LSMNAFFNNT ISSAVKQSIL AKLQEAVEAN LGTAM Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RWDD4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RWDD4
Gene Accession No. Q569B7, Q6NW29, Q9CPR1
Gene Alias BC016198; FAM28A; family with sequence similarity 28, member A; Gm1942; Protein FAM28A; RWD domain containing 4; RWD domain containing 4A; RWD domain-containing protein 4; RWD domain-containing protein 4A; Rwdd4; Rwdd4a
Gene Symbols RWDD4, Rwdd4a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RWDD4. Recombinant protein control fragment (Product #RP-96767).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192174, 201965, 502084
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.