Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RWDD4A Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297777AF647
Description
RWDD4A Polyclonal specifically detects RWDD4A in Human samples. It is validated for Western Blot.Specifications
RWDD4A | |
Polyclonal | |
Western Blot | |
FAM28A, member A, MGC10198, Protein FAM28A, RWD domain containing 4, RWD domain containing 4A, RWD domain-containing protein 4, RWD domain-containing protein 4A, RWDD4A | |
Produced in rabbits immunized with E. coli-derived Human RWDD4A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
201965 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction