Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ RX 821002 hydrochloride
GREENER_CHOICESDP

Catalog No. 132450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-245-0 50 mg
13-241-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-245-0 Supplier R&D Systems™ Supplier No. 1324/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective α2D antagonist

Potent, selective α2-adrenoceptor antagonist with very low affinity for imidazoline sites. Displays selectivity for the α2D over the α2A subtypes (pKd values are 9.7 and 8.2 respectively).

Chemical Identifiers

CAS 109544-45-8
Molecular Formula C12H15ClN2O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 270.72
Purity >99%
Molecular Weight (g/mol) 270.713
Chemical Name or Material RX 821002 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.