Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116431
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116431 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116431 Supplier Invitrogen™ Supplier No. PA5116431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RX2 Polyclonal Antibody for Western Blot

Plays a critical role in eye formation by regulating the initial specification of retinal cells and/or their subsequent proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RX2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene rx2
Gene Accession No. O42357
Gene Alias retinal homeobox gene 2; retinal homeobox protein; retinal homeobox protein Rx2; rx2; rx2 protein
Gene Symbols rx2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 51-84 amino acids from the N-terminal region of DANRE rx2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30473
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.