Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RXFP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102246 Supplier Invitrogen™ Supplier No. PA5102246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RXFP2.

The activity of this relaxin receptor is mediated by G proteins leading to stimulation of adenylate cyclase and an increase of cAMP and may also be a receptor for Leydig insulin-like peptide. Expressed in embryonic and adult gonads of males and females, as well in male gubernarculum and in brain. Not detected in kidney, spleen and heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXFP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RXFP2
Gene Accession No. Q8WXD0, Q91ZZ5
Gene Alias G protein coupled receptor affecting testicular descent; G protein-coupled receptor 106; Gpcr; GPR106; G-protein coupled receptor 106; G-protein coupled receptor affecting testicular descent; GREAT; INSL3 receptor; INSL3R; leucine-rich repeat-containing G protein-coupled receptor 8; leucine-rich repeat-containing G-protein coupled receptor 8; Lgr8; LGR8.1; Relaxin family peptide receptor 2; relaxin receptor 2; relaxin/insulin like family peptide receptor 2; relaxin/insulin-like family peptide receptor 2; Rxfp2; RXFPR2
Gene Symbols RXFP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human RXFP2(Accession Q8WXD0), corresponding to amino acid residues E121-N171.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122042, 140498, 363866
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.