Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RXFP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534174
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534174 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534174 Supplier Invitrogen™ Supplier No. PA534174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset, Horse (100%) Monkey, Panda (93%) Dog, Pig, Xenopus (86%).

The activity of this relaxin receptor is mediated by G proteins leading to stimulation of adenylate cyclase and an increase of cAMP and may also be a receptor for Leydig insulin-like peptide. Expressed in embryonic and adult gonads of males and females, as well in male gubernarculum and in brain. Not detected in kidney, spleen and heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXFP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene RXFP2
Gene Accession No. Q8WXD0, Q91ZZ5
Gene Alias G protein coupled receptor affecting testicular descent; G protein-coupled receptor 106; Gpcr; GPR106; G-protein coupled receptor 106; G-protein coupled receptor affecting testicular descent; GREAT; INSL3 receptor; INSL3R; leucine-rich repeat-containing G protein-coupled receptor 8; leucine-rich repeat-containing G-protein coupled receptor 8; Lgr8; LGR8.1; Relaxin family peptide receptor 2; relaxin receptor 2; relaxin/insulin like family peptide receptor 2; relaxin/insulin-like family peptide receptor 2; Rxfp2; RXFPR2
Gene Symbols RXFP2
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from 2nd cytoplasmic domain of human RXFP2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100033993, 122042, 140498
Target Species Human, Mouse, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.