Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RXFP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550590 Supplier Invitrogen™ Supplier No. PA550590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total Relaxin-3 receptor 1 / RXFP3 protein.

RXFP3 (Relaxin Family Peptide Receptor 3) is a Protein Coding gene. Diseases associated with RXFP3 include Schizotypal Personality Disorder and Chromosome 5P13 Duplication Syndrome. Among its related pathways are Signaling by GPCR and GPCRs, Other. Gene Ontology (GO) annotations related to this gene include G protein-coupled receptor activity and G protein-coupled peptide receptor activity. An important paralog of this gene is GPR132.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXFP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RXFP3
Gene Accession No. Q8BGE9, Q9NSD7
Gene Alias BC053073; g protein-coupled receptor SALPR; G protein-coupled receptor SALPR homolog; GPCR135; G-protein coupled receptor GPCR135; G-protein coupled receptor SALPR; MGC141998; MGC142000; relaxin 3 receptor 1; relaxin family peptide receptor 3; relaxin/insulin like family peptide receptor 3; relaxin/insulin-like family peptide receptor 3; Relaxin-3 receptor 1; relaxin-3/INSL7 receptor 1; RGD1311550; RLN3 receptor 1; Rln3r1; Rxfp3; RXFPR3; SALPR; somatostatin- and angiotensin-like peptide receptor; somatostatin and angiotensin-like peptide receptor
Gene Symbols RXFP3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human Relaxin/insulin-like family peptide receptor 3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239336, 51289
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.