Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RXFP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5141162
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5141162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5141162 Supplier Invitrogen™ Supplier No. PA5141162
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RXFP3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

RXFP3 (Relaxin Family Peptide Receptor 3) is a Protein Coding gene. Diseases associated with RXFP3 include Schizotypal Personality Disorder and Chromosome 5P13 Duplication Syndrome. Among its related pathways are Signaling by GPCR and GPCRs, Other. Gene Ontology (GO) annotations related to this gene include G protein-coupled receptor activity and G protein-coupled peptide receptor activity. An important paralog of this gene is GPR132.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXFP3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RXFP3
Gene Accession No. Q8BGE9
Gene Alias BC053073; g protein-coupled receptor SALPR; G protein-coupled receptor SALPR homolog; GPCR135; G-protein coupled receptor GPCR135; G-protein coupled receptor SALPR; MGC141998; MGC142000; relaxin 3 receptor 1; relaxin family peptide receptor 3; relaxin/insulin like family peptide receptor 3; relaxin/insulin-like family peptide receptor 3; Relaxin-3 receptor 1; relaxin-3/INSL7 receptor 1; RGD1311550; RLN3 receptor 1; Rln3r1; Rxfp3; RXFPR3; SALPR; somatostatin- and angiotensin-like peptide receptor; somatostatin and angiotensin-like peptide receptor
Gene Symbols RXFP3
Host Species Rabbit
Immunogen Synthetic peptide within Mouse Relaxin 3 receptor 1 aa 300-352.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239336
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.