Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXFP3/RLN3R1/SALPR Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297628AF750
Description
RXFP3/RLN3R1/SALPR Polyclonal specifically detects RXFP3/RLN3R1/SALPR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RXFP3/RLN3R1/SALPR | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
G protein-coupled receptor SALPR, GPCR135MGC141998, G-protein coupled receptor SALPR, relaxin 3 receptor 1, Relaxin family peptide receptor 3RLN3 receptor 1, relaxin/insulin-like family peptide receptor 3, relaxin-3 receptor 1, RLN3R1MGC142000, RXFPR3, SALPRG-protein coupled receptor GPCR135, somatostatin and angiotensin-like peptide receptor, Somatostatin- and angiotensin-like peptide receptor | |
Produced in rabbits immunized with E. coli-derived Human RXFP3/RLN3R1/SALPR fragment. | |
0.1 mL | |
GPCR | |
51289 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction