Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RXFP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102058 Supplier Invitrogen™ Supplier No. PA5102058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR100.

GPR100 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) and acts as a high affinity receptor for INSL5. Also acts as receptor for RLN3/relaxin-3, as well as bradykinin and kallidin. Binding of the ligand inhibits cAMP accumulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXFP4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RXFP4
Gene Accession No. Q8TDU9
Gene Alias G protein-coupled receptor 100; GPCR142; GPR100; G-protein coupled receptor 100; G-protein coupled receptor GPCR142; Insulin-like peptide INSL5 receptor; JEG62; MGC126556; MGC126558; relaxin 3 receptor 2; relaxin family peptide receptor 4; relaxin/insulin like family peptide receptor 4; relaxin/insulin-like family peptide receptor 4; Relaxin-3 receptor 2; relaxin-3/INSL7 receptor 2; RLN3 receptor 2; Rln3r2; Rxfp4; RXFPR4; SALPR
Gene Symbols RXFP4
Host Species Rabbit
Immunogen A synthesized peptide derived from human RXFP4(Accession Q8TDU9), corresponding to amino acid residues Q340-G374.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339403
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.