Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXR alpha/NR2B1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1001466
Description
RXR alpha/NR2B1 Polyclonal specifically detects RXR alpha/NR2B1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
RXR alpha/NR2B1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FLJ00280, FLJ00318, FLJ16020, MGC102720, NR2B1FLJ16733, Nuclear receptor subfamily 2 group B member 1, retinoic acid receptor RXR-alpha, Retinoid X receptor alpha, retinoid X receptor, alpha | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize an epitope corresponding to aa 14-28 of human RXR alpha protein and does not cross-react with either RXR beta or gamma. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/mL, Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:32000 | |
NP_002948. | |
RXRA | |
Peptide with sequence CQVNSSLTSPTGRGSM corresponding to internal region according to NP_002948.1. | |
0.1 mg | |
Cancer, Lipid and Metabolism | |
6256 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction