Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXR alpha/NR2B1 Rabbit anti-Human, Mouse, Rat, Clone: 6Y10M1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | RXR alpha/NR2B1 |
---|---|
Clone | 6Y10M1 |
Applications | ChIP Assay, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
RXR alpha/NR2B1 Monoclonal antibody specifically detects RXR alpha/NR2B1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunofluorescence, ImmunoprecipitationSpecifications
RXR alpha/NR2B1 | |
ChIP Assay, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6256 | |
IgG | |
Affinity purified |
6Y10M1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ00280, FLJ00318, FLJ16020, MGC102720, NR2B1FLJ16733, Nuclear receptor subfamily 2 group B member 1, retinoic acid receptor RXR-alpha, Retinoid X receptor alpha, retinoid X receptor, alpha | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RXR alpha/NR2B1 (P19793). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title