Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXR beta/NR2B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RXR beta/NR2B2 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RXR beta/NR2B2 Polyclonal specifically detects RXR beta/NR2B2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RXR beta/NR2B2 | |
Polyclonal | |
Rabbit | |
Cancer, Neuroscience, Transcription Factors and Regulators, Vision | |
DAUDI6, H-2RIIBP, MHC class I promoter binding protein, NR2B2MGC1831, Nuclear receptor subfamily 2 group B member 2, RCoR-1, retinoic acid receptor RXR-beta, Retinoid X receptor beta, retinoid X receptor, beta | |
RXRB | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
6257 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VAGGEQQTPEPEPGEAGRDGMGDSGRDSRSP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title