Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RXR beta/NR2B2 Antibody (PCRP-RXRB-2B6), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB281841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB281841 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB281841 Supplier Novus Biologicals Supplier No. NBP308445MFV500
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RXR beta/NR2B2 Monoclonal antibody specifically detects RXR beta/NR2B2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen RXR beta/NR2B2
Applications Flow Cytometry
Classification Monoclonal
Clone PCRP-RXRB-2B6
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias DAUDI6, H-2RIIBP, MHC class I promoter binding protein, NR2B2MGC1831, Nuclear receptor subfamily 2 group B member 2, RCoR-1, retinoic acid receptor RXR-beta, Retinoid X receptor beta, retinoid X receptor, beta
Host Species Mouse
Immunogen Recombinant full-length human RXR beta/NR2B2 protein (Uniprot: P28702)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6257
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.