Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RXR-gamma Mouse anti-Human, Clone: 12RX-2D3, Active Motif™
SDP

Catalog No. 501993085
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-3085 Supplier Active Motif Supplier No. 61033
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells (Joshi et al., 2005). The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer (Abu et al., 2005). RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors (Lane and Bailey, 2005).

Specifications

Antigen RXR-gamma
Applications Gel Shift, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 12RX-2D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene RXRG
Gene Accession No. P48443
Gene Alias Nr2b3; Nuclear receptor subfamily 2 group B member 3; Retinoic acid receptor RXR-gamma; retinoic X receptor gamma-1; retinoid X receptor; Retinoid X receptor gamma; retinoid X receptor, gamma; RXR gamma; RXR gamma-1; RXRC; Rxrg
Gene Symbols RXRG
Host Species Mouse
Immunogen This RXR-gamma antibody was raised against a peptide within the D-region of mouse RXR-gamma.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6258
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.