Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXR gamma/NR2B3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298864AF405
Description
RXR gamma/NR2B3 Polyclonal specifically detects RXR gamma/NR2B3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RXR gamma/NR2B3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
NR2B3retinoid X receptor-gamma, Nuclear receptor subfamily 2 group B member 3, retinoic acid receptor RXR-gamma, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human RXR gamma/NR2B3. | |
0.1 mL | |
Cancer, Transcription Factors and Regulators | |
6258 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction