Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXR gamma/NR2B3 Antibody (PCRP-RXRG-5H4), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314170C
This item is not returnable.
View return policy
Description
RXR gamma/NR2B3 Monoclonal antibody specifically detects RXR gamma/NR2B3 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
RXR gamma/NR2B3 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-RXRG-5H4 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
NR2B3retinoid X receptor-gamma, Nuclear receptor subfamily 2 group B member 3, retinoic acid receptor RXR-gamma, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC | |
Recombinant full-length human RXR gamma/NR2B3 protein (Uniprot: P48443) | |
0.1 mL | |
Cancer, Transcription Factors and Regulators | |
6258 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction