Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXR gamma/NR2B3 Mouse anti-Human, Clone: PCRP-RXRG-5H4, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | RXR gamma/NR2B3 |
---|---|
Clone | PCRP-RXRG-5H4 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
Applications | Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array |
Description
RXR gamma/NR2B3 Monoclonal antibody specifically detects RXR gamma/NR2B3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
RXR gamma/NR2B3 | |
0.2 mg/ml | |
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
NR2B3retinoid X receptor-gamma, Nuclear receptor subfamily 2 group B member 3, retinoic acid receptor RXR-gamma, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC | |
Recombinant full-length human RXR gamma/NR2B3 protein (Uniprot: P48443) | |
Primary | |
Store at 4°C. |
PCRP-RXRG-5H4 | |
Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P48443 | |
6258 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title