Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RXRA Monoclonal Antibody (5E7)
GREENER_CHOICE

Catalog No. PIMA549193
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549193 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549193 Supplier Invitrogen™ Supplier No. MA549193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human A549 whole cell, human HepG2 whole cell, human Caco-2 whole cell, human HL-60 whole cell, rat liver tissue, mouse liver tissue. Flow: A549 cell, CACO-2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXRA
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5E7
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Rxra
Gene Accession No. P19793, P28700, Q05343
Gene Alias 9530071D11Rik; Nr2b1; Nuclear receptor subfamily 2 group B member 1; retinoic acid receptor RXR-alpha; retinoid X nuclear receptor alpha; retinoid X receptor; retinoid X receptor alpha; retinoid X receptor, alpha; rxr alpha; RXR alpha 1; Rxra; RXRalpha1
Gene Symbols Rxra
Host Species Mouse
Immunogen E. coli-derived human RXRA recombinant protein (Position: A226-T462).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20181, 25271, 6256
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.