Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXRA Mouse anti-Human, Mouse, Clone: 4H6C4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | RXRA |
---|---|
Clone | 4H6C4 |
Concentration | 0.45 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6301
![]() ![]() |
Proteintech Group Inc
601981IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.Specifications
RXRA | |
0.45 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NR2B1, Retinoid X receptor alpha, retinoid X receptor, alpha, RXRA | |
RXRA | |
IgG1 | |
Protein G | |
Antibody |
4H6C4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P19793, P28700 | |
20181, 6256 | |
RXRA Fusion Protein Ag0987 | |
Primary | |
-20°C | |
RXRA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title