Learn More
Invitrogen™ RXRA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579945
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human COLO-320 whole cell, human A431 whole cell, human MCF-7 whole cell, rat heart tissue, mouse heart tissue. IHC: mouse liver tissue, rat liver tissue. ICC/IF: A431 cell. Flow: A431 cell.
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
Specifications
RXRA | |
Polyclonal | |
Unconjugated | |
Rxra | |
9530071D11Rik; Nr2b1; Nuclear receptor subfamily 2 group B member 1; retinoic acid receptor RXR-alpha; retinoid X nuclear receptor alpha; retinoid X receptor; retinoid X receptor alpha; retinoid X receptor, alpha; rxr alpha; RXR alpha 1; Rxra; RXRalpha1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20181, 25271, 6256 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P19793, P28700, Q05343 | |
Rxra | |
E. coli-derived human RXRA recombinant protein (Position: A226-T462). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.