Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RXRB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1815 Supplier Invitrogen™ Supplier No. PA1815
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-815 detects retinoid X receptor (RXR) beta from human and mouse cells as well as transfected human and mouse RXR beta. PA1-815 has been successfully used in Western blot, immunoprecipitation and gel shift procedures. By Western blot, this antibody detects an ~50 kDa protein representing RXR beta from COS cells transfected with the human gene. Gel shift assays reveal supershifting of vitamin D receptor (VDR)/RXR beta/vitamin D receptor response element (VDRE) complexes when PA1-815 is preincubated with RXR beta prior to addition of VDR. The PA1-815 immunogen is a synthetic peptide corresponding to residues Q(190) K S D Q G V E G P G A T(202) of mouse RXR beta. Recombinant human VDR (Cat. # RP-710) and human RXR beta (catalog # RP-812) proteins can be purchased separately for control experiments in Western blot, immunoprecipitation and gel shift procedures.

This gene encodes a member of the retinoid X receptor (RXR) family of nuclear receptors which are involved in mediating the effects of retinoic acid (RA). This receptor forms homodimers with the retinoic acid, thyroid hormone, and vitamin D receptors, increasing both DNA binding and transcriptional function on their respective response elements. The gene lies within the major histocompatibility complex (MHC) class II region on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXRB
Applications Gel Shift, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene Rxrb
Gene Accession No. P28702, P28704
Gene Alias AL023085; DADB-100D22.1; DAUDI6; H-2RIIBP; MGC1831; MHC class I promoter binding protein; MHC class I regulatory element-binding protein H-2RIIBP; NR2B2; nuclear receptor coregulator 1; nuclear receptor co-regulator 1; nuclear receptor subfamily 2 group B member 2; putative retinoid X receptor beta; RCoR-1; Retinoic acid receptor RXR-beta; retinoic acid receptor, beta; retinoid X receptor; retinoid X receptor beta; retinoid X receptor, beta; Rub; RXR Beta; Rxrb; RXR-beta
Gene Symbols Rxrb
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues Q(190) K S D Q G V E G P G A T(202) of mouse RXR beta.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20182, 6257
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.