Learn More
RXRB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521610
Description
PA5-21610 targets Retinoid X Receptor beta in WB applications and shows reactivity with Human samples. The PA5-21610 immunogen is a synthetic peptide corresponding to a region within amino acids 20 and 113 of Retinoid X Receptor beta (Uniprot IDNo.P28702). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
| RXRB | |
| Polyclonal | |
| Unconjugated | |
| P28702 | |
| RXRB | |
| Synthetic peptide corresponding to a region within amino acids 20 and 113 of Retinoid X Receptor beta (Uniprot IDNo.P28702) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| RXRB | |
| DADB-100D22.1, DAUDI6, H-2RIIBP, NR2B2, RCoR-1, MHC class I promoter binding protein, nuclear receptor subfamily 2 group B member 2, retinoic acid receptor RXR-beta, retinoid X nuclear receptor beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6257 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.