Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RXRG Mouse anti-Human, Clone: 1373, Abnova™

Catalog No. 89035552 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-552 Supplier Abnova Corporation Supplier No. MAB14154

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human RXRG.

  • High specificity
  • High reactivity

Specifications

Antigen RXRG
Applications Western Blot
Classification Monoclonal
Clone 1373
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human RXRG.
Dilution Western Blot The optimal working dilution should be determined by the end user.
Formulation In HEPES, 0.15 M NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
Gene Accession No. P48443
Gene Alias NR2B3/RXRC
Gene Symbols RXRG
Host Species Mouse
Immunogen A hinge peptide corresponding to human RXRG.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6258
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.