Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RXRG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581256
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human RXRG.
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells (Joshi et al., 2005). The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer (Abu et al., 2005). RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors (Lane and Bailey, 2005).
Specifications
RXRG | |
Polyclonal | |
Unconjugated | |
RXRG | |
Nr2b3; Nuclear receptor subfamily 2 group B member 3; retinoic acid receptor RXR-gamma; retinoic X receptor gamma-1; retinoid X receptor; retinoid X receptor gamma; retinoid X receptor, gamma; RXR gamma; RXR gamma-1; RXRC; RXRG | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
6258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
P48443 | |
RXRG | |
A synthetic peptide corresponding to the center region of the Human RXRG. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction