Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RY586 Mouse Anti-Mouse H-2Kb/SIINFEKL, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570018
Description
H-2Kb/SIINFEKL Monoclonal antibody specifically detects H-2Kb/SIINFEKL in Mouse samples. It is validated for Flow cytometry
Specifications
H-2Kb/SIINFEKL | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
25-D1.16.rMAb (also known as 25-D1.16) | |
RY586 | |
H-2Kb/SIINFEKL; H-2Kb:SIINFEKL; SIINFEKL-bound H-2Kb | |
SIINFEKL-pulsed RMA-S cells | |
0.1 mg | |
14972 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction