Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RY586 Rat Anti-Mouse CD366 (TIM-3), BD Horizon™
Rat Monoclonal Antibody
$104.00 - $262.00
Specifications
Antigen | CD366 (TIM-3) |
---|---|
Clone | 25F.1D6 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD366 (TIM-3) Monoclonal antibody specifically detects CD366 (TIM-3) in Mouse samples. It is validated for Flow cytometrySpecifications
CD366 (TIM-3) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
171285 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
25F.1D6 | |
Flow Cytometry | |
RY586 | |
Rat | |
Mouse | |
Cd366; TIM-3; Tim3; TIMD-3; Timd3; HAVcr-2; Havcr2 | |
AE7 Th1 Cells and Th1-like Cells from TCR Transgenic Mice | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title