Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ryanodine Receptor 2 Antibody (C3-33) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280143
Description
Ryanodine Receptor 2 Monoclonal specifically detects Ryanodine Receptor 2 in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Chicken, Fish, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Ryanodine Receptor 2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1 ug/ml, Flow Cytometry 1:10-1:000, Immunohistochemistry 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1 ug/ml, Immunohistochemistry-Frozen 1 ug/ml | |
arrhythmogenic right ventricular dysplasia 2, ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, Cardiac muscle-type ryanodine receptor, cardiac-type ryanodine receptor, hRYR-2, islet-type ryanodine receptor, kidney-type ryanodine receptor, ryanodine receptor 2, ryanodine receptor 2 (cardiac), RyR2, RYR-2, type 2 ryanodine receptor, VTSIP | |
Mouse | |
Protein G purified | |
RUO | |
6262 | |
Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Chicken, Fish, Guinea Pig, Rabbit | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
C3-33 | |
Unconjugated | |
PBS | |
RYR2 | |
Canine cardiac ryanodine receptor (RyR2) | |
100 μg | |
Primary | |
Detects Ryanodine Receptor 2. Also shows cross-reactivity with Ryanodine Receptor 1 in amphibian, canine, chicken, fish, guinea pig and rat tissues. | |
Store at -20°C. Avoid freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction