Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ryanodine
GREENER_CHOICESDP

Catalog No. 13291 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-291 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-291 Supplier Tocris Bioscience™ Supplier No. 1329/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ca2+ release inhibitor

Potent inhibitor of Ca2+ release from sarcoplasmic reticulum (IC50 values are 10 and 2.8 nM in skeletal and cardiac muscle respectively).

Chemical Identifiers

CAS 15662-33-6
Molecular Formula C25H35NO9

Specifications

Quantity 1 mg
Formula Weight 493.55
Purity >98%
Molecular Weight (g/mol) 493.553
Chemical Name or Material Ryanodine
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.