Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RYBP/DEDAF Antibody (OTI2B4), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273962UV
Description
RYBP/DEDAF Monoclonal specifically detects RYBP/DEDAF in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot.Specifications
RYBP/DEDAF | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RYBP | |
Full length recombinant protein of human RYBP/DEDAF (NP_036366) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Canine, Monkey | |
IgG1 |
Western Blot | |
OTI2B4 | |
Western Blot | |
AAP1, APAP-1, Apoptin-associating protein 1, Death effector domain-associated factor, DEDAFYY1 and E4TF1-associated factor 1, RING1 and YY1 binding protein, RING1 and YY1-binding protein, ring1 interactor RYBP, YEAF1DED-associated factor, YY1 and E4TF1 associated factor 1 | |
Mouse | |
Affinity Purified | |
Apoptosis | |
23429.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction