Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RYBP/DEDAF Antibody (SR1538), Novus Biologicals™
SDP

Catalog No. p-200006487 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259605 100 μg
NB258539 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259605 Supplier Novus Biologicals Supplier No. NBP321976100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RYBP/DEDAF Monoclonal antibody specifically detects RYBP/DEDAF in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen RYBP/DEDAF
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SR1538
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias AAP1, APAP-1, Apoptin-associating protein 1, Death effector domain-associated factor, DEDAFYY1 and E4TF1-associated factor 1, RING1 and YY1 binding protein, RING1 and YY1-binding protein, ring1 interactor RYBP, YEAF1DED-associated factor, YY1 and E4TF1 associated factor 1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RYBP/DEDAF (Uniprot #: Q8N488)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 23429
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.