Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RYBP Monoclonal Antibody (OTI2D4), TrueMAB™, OriGene
SDP

Catalog No. 502540193
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0193 30 μL
50-167-5298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0193 Supplier OriGene Supplier No. TA504652S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RYBP Monoclonal antibody specifically detects RYBP in Canine, Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen RYBP
Applications Western Blot
Classification Monoclonal
Clone OTI2D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Rybp
Gene Accession No. Q8CCI5, Q8N488
Gene Alias 2410018J24Rik; AAP1; APAP-1; apoptin-associating protein 1; Death effector domain-associated factor; DEDAF; DED-associated factor; RING1 and YY1 binding protein; RING1 and YY1-binding protein; ring1 interactor RYBP; Rybp; YEAF1; YY1 and E4TF1 associated factor 1; YY1 and E4TF1-associated factor 1
Host Species Mouse
Immunogen Full length human recombinant protein of human RYBP produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23429, 484690, 56353
Target Species Canine, Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.