Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RYBP Mouse anti-Canine, Human, Mouse, Clone: OTI2B4, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA504651

Description
RYBP involves component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1-like complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A ′Lys-119′, rendering chromatin heritably changed in its expressibility. Component of a PRC1-like complex that mediates monoubiquitination of histone H2A ′Lys-119′ on the X chromosome and is required for normal silencing of one copy of the X chromosome in XX females. May stimulate ubiquitination of histone H2A ′Lys-119′ by recruiting the complex to target sites. Inhibits ubiquitination and subsequent degradation of TP53, and thereby plays a role in regulating transcription of TP53 target genes. May also regulate the ubiquitin-mediated proteasomal degradation of other proteins like FANK1 to regulate apoptosis. May be implicated in the regulation of the transcription as a repressor of the transcriptional activity of E4TF1. May bind to DNA. May play a role in the repression of tumor growth and metastasis in breast cancer by down-regulating SRRM3.Specifications
RYBP | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8CCI5, Q8N488 | |
RYBP | |
Full length human recombit protein of human RYBP produced in E.coli. | |
100 μL | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
OTI2B4 | |
Unconjugated | |
RYBP | |
AAP1, APAP-1, DEDAF, YEAF1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23429, 484690, 56353 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction