Learn More
Invitrogen™ RYK Polyclonal Antibody

Description
Immunogen sequence: LWELMTLGQT PYVDIDPFEM AAYLKDGYRI AQPINCPDEL FAVMACCWAL DPEERPKFQQ LVQCLTEFHA ALG Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | RYK |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RYK |
| Gene Accession No. | P34925 |
| Gene Alias | AW536699; D3S3195; ERK3; ERK-3; growth factor receptor; hydroxyaryl-protein kinase; JTK5; JTK5A; JTK5A protein tyrosine kinase; kinase VIK; Met-related kinase; Mrk; NYK-R; receptor-like tyrosine kinase; RYK; RYK1; tyrosine-protein kinase RYK; Vik |
| Gene Symbols | RYK |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.