Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RyR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109277 Supplier Invitrogen™ Supplier No. PA5109277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Human fetal skeletal muscle tissue, mouse cerebellum tissue, SiHa.

Ryanodine receptors have been shown to play critical roles in the intracellular Ca2+ signaling occurring during cell activation in muscle cells and non-muscled cells. Ryanodine receptor, ~500 kDa, can be isolated from a variety of cell types such as lymphocytes, macrophages, granulocytes, fibroblasts, epithelial, endothelial cells, skeletal muscles, cardiac muscle and brain tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RyR1
Applications Dot Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene RYR1
Gene Accession No. E9PZQ0, F1LMY4, P21817
Gene Alias AI528790; calcium release channel; calcium release channel (ryanodine receptor 1); calcium release channel protein; CCO; central core disease of muscle; CRC; CRC1, RYR1; halothane; LOW QUALITY PROTEIN: ryanodine receptor 1; MHS; MHS1; porcine stress syndrome; PPP1R137; protein phosphatase 1, regulatory subunit 137; ryanodine receptor; ryanodine receptor 1; ryanodine receptor 1 (skeletal); ryanodine receptor 1, skeletal muscle; ryanodine receptor 1-like, skeletal muscle; RYDR; Ryr; Ryr1; RYR-1; Ryr1l; sarcoplasmic reticulum calcium release channel; skeletal muscle calcium release channel; Skeletal muscle ryanodine receptor; skeletal muscle-type ryanodine receptor; skrr; type 1 ryanodine receptor; type 1-like ryanodine receptor; unnamed protein product
Gene Symbols RYR1
Host Species Rabbit
Immunogen Synthetic peptide within Human RYR1 aa 1,354-1,403.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114207, 20190, 6261
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.