Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ryuvidine
GREENER_CHOICE

Catalog No. 260910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-091-0 10 mg
26-095-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-091-0 Supplier Tocris Bioscience™ Supplier No. 2609/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

SETD8 inhibitor; also CDK4 inhibitor

Inhibitor of SETD8 protein lysine methyltransferase (PKMT) (IC50 = 0.5 μM); suppresses H4K20 monomethylation in vitro. Also inhibits cyclin-dependent kinase (CDK) 4 (IC50 = 6.0 μM at CDK4/cyclin D1). Induces S phase accumulation in HEK293T cells. Cytotoxic against a range of human cancer cells.

Chemical Identifiers

CAS 265312-55-8
Molecular Formula C15H12N2O2S

Specifications

Quantity 10 mg
Formula Weight 284.33
Purity >98%
Molecular Weight (g/mol) 284.333
Chemical Name or Material Ryuvidine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.