Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ S 07662

Catalog No. 518810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-881-0 10 mg
51-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-881-0 Supplier Tocris Bioscience™ Supplier No. 5188/10
This item is not returnable. View return policy

Inverse agonist of the constitutive androstane receptor (CAR)

S 07662 is an inverse agonist of the constitutive androstane receptor (CAR). Inhibits CAR activity via recruitment of nuclear receptor co-repressor (NCoR). Suppresses CITCO -induced CYP2B6 gene expression in human primary hepatocytes.

Specifications

Purity >98%
Quantity 10 mg
Product Type S 07662
Molecular Weight (g/mol) 300.38
Inhibitors S 07662

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.