Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1,1'-Bi-2-naphthol, 99%

Catalog No. AC224311000
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC224311000 100 mg
AC224310010 1 g
AC224310050 5 g
AC224310250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC224311000 Supplier Thermo Scientific Chemicals Supplier No. 224311000

CAS: 18531-99-2 | C20H14O2 | 286.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18531-99-2
InChI Key PPTXVXKCQZKFBN-UHFFFAOYSA-N
Synonym 1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
PubChem CID 11762
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 205°C to 211°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C20H14O2
Linear Formula HOC10H6C10H6OH
Quantity 100 mg
Fieser 16,132
Merck Index 15, 1230
Specific Rotation Condition −35.50° (20°C c=1,THF)
Specific Rotation -33° to -38° (20°C, 589nm) (c=1, THF)
Molecular Weight (g/mol) 286.33
Formula Weight 286.32
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(-)-1, 1'-Bi-2-naphthol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.