Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(1-aminopropyl)phosphonic acid, 99%, Thermo Scientific™

Catalog No. ac376435000
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98048-99-8
Molecular Formula C3H10NO3P
Molecular Weight (g/mol) 139.091
MDL Number MFCD00799472
InChI Key DELJNDWGTWHHFA-VKHMYHEASA-N
Synonym 1s-+-1-aminopropyl phosphonic acid, unii-wnd449nj8r, wnd449nj8r, s-1-aminopropyl phosphonic acid, 1s-1-aminopropylphosphonic acid, ampropylfos, s, ampropylfos, +, 1s-1-aminopropyl phosphonic acid
PubChem CID 6994862
IUPAC Name [(1S)-1-aminopropyl]phosphonic acid
SMILES CCC(N)P(=O)(O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3CH2CH(NH2)P(O)(OH)2
Quantity 500 mg
Specific Rotation Condition + 19.00 (20.00°C c2.0,1N NaOH)
Specific Rotation +16° to +19° (20°C, 589nm) (C=2, 1N NaOH)
Formula Weight 139.09
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material (S)-(1-Aminopropyl)phosphonic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.