Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Bromo-2-methylbutane, 97%, stab. with potassium carbonate

Catalog No. aah2707806
Click to view available options
Quantity:
5 g
25 g

CAS: 534-00-9 | C5H11Br | 151.05 g/mol

(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-00-9
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00078662
InChI Key XKVLZBNEPALHIO-YFKPBYRVSA-N
Synonym s-1-bromo-2-methylbutane, s-+-1-bromo-2-methylbutane, butane, 1-bromo-2-methyl-, 2s, unii-cgs982pbtw, 2s-1-bromo-2-methylbutane, d-amyl bromide, cgs982pbtw, 2s-1-bromo-2-methyl-butane, +-2-methylbutyl bromide, butane, 1-bromo-2-methyl-, s
PubChem CID 5464167
IUPAC Name (2S)-1-bromo-2-methylbutane
SMILES CC[C@H](C)CBr

Specifications

Density 1.223
Boiling Point 121°C to 122°C
Flash Point 22°C (79°F)
Refractive Index 1.4455
Quantity 5 g
UN Number UN1993
Beilstein 1718815
Sensitivity Light sensitive; May form a precipitate
Merck Index 14,601
Solubility Information Insoluble in water.
Optical Rotation +4.5° (Neat)
Formula Weight 151.05
Percent Purity 97%
Chemical Name or Material (S)-(+)-1-Bromo-2-methylbutane, stabilized with potassium carbonate
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 208-583-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.