Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene 97.0+%, TCI America™
SDP

Catalog No. D38221G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 163169-29-7
Molecular Formula C28H28FeNOP
Molecular Weight (g/mol) 481.36
MDL Number MFCD15144869
InChI Key FELCHLOAEMNOQM-KISGOYOSNA-N
Synonym (S,S)-ip-FOXAP
IUPAC Name (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine
SMILES [Fe].c1cccc1.CC(C)[C@H]1COC(=N1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 160°C
Quantity 1 g
Formula Weight 481.36
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.