Learn More
S-100 Monoclonal Antibody (4C4.9), Zeta
Mouse Monoclonal Antibody Cocktail
$274.38
Specifications
| Antigen | S-100 |
|---|---|
| Clone | 4C4.9 |
| Concentration | 200 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-221-5836
|
ZETA CORPORATION
Z2055MS |
500 μL |
Each of 1 for $274.38
|
|
|||||
Description
A recommended positive control tissue for this product is Melanoma, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
S-100 Monoclonal antibody specifically detects S-100 in Bovine, Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| S-100 | |
| 200 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| tris with BSA, NP-40 and <0.1% sodium azide | |
| AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P | |
| Purified bovine brain S100 p | |
| Primary | |
| 2-8°C | |
| S100A1 |
| 4C4.9 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Bovine, Human | |
| P02638, P02639, P04271, P23297 | |
| 525716, 528735, 6271, 6285 | |
| IgG2a κ | |
| Protein A | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.