Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Acetoxypropionic Acid 98.0+%, TCI America™
SDP

Catalog No. A129925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A129925G 25 g
A12995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A129925G Supplier TCI America Supplier No. A129925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6034-46-4
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00191370
InChI Key WTLNOANVTIKPEE-VKHMYHEASA-N
Synonym (-)-O-Acetyl-L-lactic Acid
PubChem CID 6326324
IUPAC Name (2S)-2-acetyloxypropanoic acid
SMILES CC(C(=O)O)OC(=O)C

Specifications

Color Yellow
Boiling Point 142°C
Quantity 25 g
Formula Weight 132.12
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Acetoxypropionic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.